missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 10 Antibody (18B6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26178-100ul
This item is not returnable.
View return policy
Description
Cytokeratin 10 Monoclonal antibody specifically detects Cytokeratin 10 in Human samples. It is validated for ELISA, Immunohistochemistry
Specifications
| Cytokeratin 10 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry | |
| 18B6 | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| BCIE, BIE, CK10, CK-10, cytokeratin 10, Cytokeratin-10, EHK, K10keratosis palmaris et plantaris, keratin 10, Keratin 10 (epidermolytic hyperkeratosis, keratosis palmaris et plantaris), keratin, type I cytoskeletal 10, keratin-10, KPP | |
| A synthesized peptide derived from Human Cytokeratin 10 [UniProt P13645] | |
| 100 μL | |
| Cell Biology, Cellular Markers | |
| 3858 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction