missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytohesin 4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-56913
This item is not returnable.
View return policy
Description
Cytohesin 4 Polyclonal specifically detects Cytohesin 4 in Human samples. It is validated for Western Blot.
Specifications
| Cytohesin 4 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| CYT4PH, SEC7 and coiled-coil domain-containing protein 4, cytohesin 4, cytohesin-4, pleckstrin homology, Sec7 and coiled/coil domains 4, pleckstrin homology, Sec7 and coiled-coil domains 4, PSCD4DJ63G5.1 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 27128 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9UIA0 | |
| CYTH4 | |
| Synthetic peptides corresponding to PSCD4(pleckstrin homology, Sec7 and coiled-coil domains 4) The peptide sequence was selected from the N terminal of PSCD4. Peptide sequence NKTAIGTYLGERDPINLQVLQAFVDCHEFANLNLVQALRQFLWSFRLPGE. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Pig: 100%; Bovine: 100%; Rat: 92%; Mouse: 92%; Western clawed frog: 90%; Xenopus: 90%; Zebrafish: 90%; Rainbow smelt: 90%; Atlantic salmon: 78%; Green puffer: 78%; Chicken: 78%; Green monkey: 78%; Human: 100%;. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction