missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytohesin 2 Antibody (CYT2-21) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£175.00
Specifications
| Antigen | Cytohesin 2 |
|---|---|
| Clone | CYT2-21 |
| Concentration | 2 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
| Applications | Western Blot |
Description
Cytohesin 2 Monoclonal specifically detects Cytohesin 2 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Cytohesin 2 | |
| 2 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Membrane Trafficking and Chaperones | |
| ARF exchange factor, ARF nucleotide-binding site opener, ARNOPH, SEC7 and coiled-coil domain-containing protein 2, CTS18.1, cytohesin 2, pleckstrin homology, Sec7 and coiled-coil domains 2 (cytohesin-2), Protein ARNO, PSCD2LCTS18, PSCD2pleckstrin homology, Sec7 and coiled-coil domains 2, Sec7 and coiled/coil domains 2 (cytohesin-2) | |
| CYTH2 | |
| IgG1 | |
| Protein A or G purified | |
| does not cross-react with other members of the cytohesin family. |
| CYT2-21 | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 9266 | |
| Recombinant full length protein (Human). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title