missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytohesin 2 Antibody (CYT2-21) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-12421-0.025ml
This item is not returnable.
View return policy
Description
Cytohesin 2 Monoclonal specifically detects Cytohesin 2 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Cytohesin 2 | |
| Monoclonal | |
| 2 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| CYTH2 | |
| Recombinant full length protein (Human). | |
| 0.025 mL | |
| Membrane Trafficking and Chaperones | |
| 9266 | |
| Human | |
| Purified |
| Western Blot | |
| CYT2-21 | |
| Unconjugated | |
| ARF exchange factor, ARF nucleotide-binding site opener, ARNOPH, SEC7 and coiled-coil domain-containing protein 2, CTS18.1, cytohesin 2, pleckstrin homology, Sec7 and coiled-coil domains 2 (cytohesin-2), Protein ARNO, PSCD2LCTS18, PSCD2pleckstrin homology, Sec7 and coiled-coil domains 2, Sec7 and coiled/coil domains 2 (cytohesin-2) | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| does not cross-react with other members of the cytohesin family. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction