missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytohesin-1 Antibody (2E11), Novus Biologicals™
Mouse Monoclonal Antibody
£155.00
Specifications
| Antigen | Cytohesin-1 |
|---|---|
| Clone | 200000000000 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1.0 μg/mL, Flow Cytometry 3 - 5 ug 106 cells, Immunocytochemistry/Immunofluorescence 1:20 |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18753014
|
Novus Biologicals
NBP2-22512-0.025mg |
0.025 mg | |||||||
Description
Cytohesin-1 Monoclonal specifically detects Cytohesin-1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
| Cytohesin-1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Membrane Trafficking and Chaperones | |
| Q15438 | |
| 9267 | |
| Purified, His-tagged cytohesin-1. | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 200000000000 | |
| Western Blot 1.0 μg/mL, Flow Cytometry 3 - 5 ug 106 cells, Immunocytochemistry/Immunofluorescence 1:20 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| B2-1, cytoadhesin 1, cytohesin 1, cytohesin-1, D17S811ECYTOHESIN-1, FLJ34050, FLJ41900, homolog of secretory protein SEC7, PH, SEC7 and coiled-coil domain-containing protein 1, pleckstrin homology, Sec7 and coiled-coil domains 1, PSCD1, SEC7, SEC7 homolog B2-1 | |
| CYTH1 | |
| IgG1 | |
| PEG purified | |
| This antibody reacts with Human. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title