missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytohesin-1 Antibody (2E11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22512-0.025mg
This item is not returnable.
View return policy
Description
Cytohesin-1 Monoclonal specifically detects Cytohesin-1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| Cytohesin-1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1.0 μg/mL, Flow Cytometry 3 - 5 ug 106 cells, Immunocytochemistry/Immunofluorescence 1:20 | |
| B2-1, cytoadhesin 1, cytohesin 1, cytohesin-1, D17S811ECYTOHESIN-1, FLJ34050, FLJ41900, homolog of secretory protein SEC7, PH, SEC7 and coiled-coil domain-containing protein 1, pleckstrin homology, Sec7 and coiled-coil domains 1, PSCD1, SEC7, SEC7 homolog B2-1 | |
| Mouse | |
| PEG purified | |
| RUO | |
| Primary | |
| This antibody reacts with Human. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| 200000000000 | |
| Unconjugated | |
| Q15438 | |
| CYTH1 | |
| Purified, His-tagged cytohesin-1. | |
| 0.025 mg | |
| Membrane Trafficking and Chaperones | |
| 9267 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction