missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Cytochrome P450 3A5 Antibody (F18 P3 B6), Novus Biologicals™
Mouse Monoclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | Cytochrome P450 3A5 |
|---|---|
| Clone | F18 P3 B6 |
| Dilution | Western Blot : 1-2 μg/mL, ELISA, Immunohistochemistry-Paraffin : 1-5 μg/mL, Immunohistochemistry-Frozen : 1-5 μg/mL |
| Applications | ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066918
|
Novus Biologicals™
NBP3-30887-200ug |
200 μg |
£386.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066839
|
Novus Biologicals™
NBP3-30887-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytochrome P450 3A5 Monoclonal antibody specifically detects Cytochrome P450 3A5 in Human samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen)Specifications
| Cytochrome P450 3A5 | |
| Western Blot : 1-2 μg/mL, ELISA, Immunohistochemistry-Paraffin : 1-5 μg/mL, Immunohistochemistry-Frozen : 1-5 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| aryl hydrocarbon hydroxylase, CP35, CYPIIIA5, cytochrome P-450, cytochrome P450 3A5, Cytochrome P450 HLp2, cytochrome P450, family 3, subfamily A, polypeptide 5, cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 5, Cytochrome P450-PCN3, DKFZp686L16231, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, niphedipine oxidase, P450PCN3FLJ31317, PCN3, xenobiotic monooxygenase | |
| Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with a synthetic peptide derived from the C-terminus of the human Cytochrome P450 3A5 protein and mouse myeloma Ag8563 cells. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| F18 P3 B6 | |
| ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Lipid and Metabolism | |
| Phosphate buffered saline | |
| 1577 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title