missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Cytochrome P450 3A5 Antibody (F18 P3 B6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-30887-200ug
This item is not returnable.
View return policy
Description
Cytochrome P450 3A5 Monoclonal antibody specifically detects Cytochrome P450 3A5 in Human samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen)
Specifications
| Cytochrome P450 3A5 | |
| Monoclonal | |
| Unconjugated | |
| Phosphate buffered saline | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| F18 P3 B6 | |
| Western Blot : 1-2 μg/mL, ELISA, Immunohistochemistry-Paraffin : 1-5 μg/mL, Immunohistochemistry-Frozen : 1-5 μg/mL | |
| aryl hydrocarbon hydroxylase, CP35, CYPIIIA5, cytochrome P-450, cytochrome P450 3A5, Cytochrome P450 HLp2, cytochrome P450, family 3, subfamily A, polypeptide 5, cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 5, Cytochrome P450-PCN3, DKFZp686L16231, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, niphedipine oxidase, P450PCN3FLJ31317, PCN3, xenobiotic monooxygenase | |
| Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with a synthetic peptide derived from the C-terminus of the human Cytochrome P450 3A5 protein and mouse myeloma Ag8563 cells. | |
| 200 μg | |
| Lipid and Metabolism | |
| 1577 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction