missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 2C8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£292.00 - £435.00
Specifications
| Antigen | Cytochrome P450 2C8 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18481002
|
Novus Biologicals
NBP1-88055-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18218027
|
Novus Biologicals
NBP1-88055 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytochrome P450 2C8 Polyclonal specifically detects Cytochrome P450 2C8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Cytochrome P450 2C8 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| CPC8, CYPIIC8, cytochrome P450 2C8, Cytochrome P450 form 1, Cytochrome P450 IIC2, Cytochrome P450 MP-12, Cytochrome P450 MP-20, cytochrome P450, family 2, subfamily C, polypeptide 8, cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 8, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, MP-12/MP-20, P450 form 1, S-mephenytoin 4-hydroxylase, xenobiotic monooxygenase | |
| CYP2C8 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 1558 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:KRICAGEGLARMELFLFLTTILQNFNLKSVDDLKNLNTTAVTKGIVSLPPSYQICFIPV | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title