missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 2A6, 2A7, 2A13 Antibody (F16 P2 D8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£169.00 - £319.00
Specifications
| Antigen | Cytochrome P450 2A6, 2A7, 2A13 |
|---|---|
| Clone | F16 P2 D8 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18698798
|
Novus Biologicals
NBP2-50184-0.025ml |
0.025 mL |
£169.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18622009
|
Novus Biologicals
NBP2-50184 |
0.1 mL |
£319.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytochrome P450 2A6, 2A7, 2A13 Monoclonal antibody specifically detects Cytochrome P450 2A6, 2A7, 2A13 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| Cytochrome P450 2A6, 2A7, 2A13 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
| PBS | |
| 1548 | |
| IgG1 κ | |
| Protein A purified |
| F16 P2 D8 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| cytochrome P450 2A13, cytochrome P450 2A6, cytochrome P450 2A7, cytochrome P450, family 2, subfamily A, polypeptide 13, cytochrome P450, family 2, subfamily A, polypeptide 6, cytochrome P450, family 2, subfamily A, polypeptide 7 | |
| ovalbumin-conjugated synthetic peptide RNYTMSFLPR. CYP2A6, CYP2A7and CYP2A13 have the same c-terminal sequence. (C-terminal sequence identical in human and rhesus macaque) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title