missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 2A6, 2A7, 2A13 Antibody (F16 P2 D8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50184-0.025ml
This item is not returnable.
View return policy
Description
Cytochrome P450 2A6, 2A7, 2A13 Monoclonal antibody specifically detects Cytochrome P450 2A6, 2A7, 2A13 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| Cytochrome P450 2A6, 2A7, 2A13 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| cytochrome P450 2A13, cytochrome P450 2A6, cytochrome P450 2A7, cytochrome P450, family 2, subfamily A, polypeptide 13, cytochrome P450, family 2, subfamily A, polypeptide 6, cytochrome P450, family 2, subfamily A, polypeptide 7 | |
| ovalbumin-conjugated synthetic peptide RNYTMSFLPR. CYP2A6, CYP2A7and CYP2A13 have the same c-terminal sequence. (C-terminal sequence identical in human and rhesus macaque) | |
| 0.025 mL | |
| Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
| 1548 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry | |
| F16 P2 D8 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction