missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 26B1 Mouse anti-Human, Mouse, Rat, Clone: T5P3G2/A3, Novus Biologicals™
Mouse Monoclonal Antibody
£173.00 - £315.00
Specifications
| Antigen | Cytochrome P450 26B1 |
|---|---|
| Clone | T5P3G2/A3 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18129927
|
Novus Biologicals
NBP2-50195-0.025ML |
0.025 mL |
£173.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18692009
|
Novus Biologicals
NBP2-50195 |
0.1 mL |
£315.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytochrome P450 26B1 Monoclonal specifically detects Cytochrome P450 26B1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Cytochrome P450 26B1 | |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| CYP26A2, Cytochrome P450 26A2, cytochrome P450 26B1, cytochrome P450 CYP26B1 variant 2, Cytochrome P450 retinoic acid-inactivating 2, cytochrome P450 retinoid metabolizing protein, cytochrome P450, family 26, subfamily B, polypeptide 1, Cytochrome P450RAI-2, DKFZp686G0638, EC 1.14, MGC129613, P450RAI2, P450RAI-2, Retinoic acid-metabolizing cytochrome | |
| CYP26B1 | |
| IgG1 κ | |
| Protein G purified |
| T5P3G2/A3 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 56603 | |
| Ovalbumin-conjugated synthetic peptide DSNQNEILPE (C-terminal sequence) | |
| Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title