missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 26B1 Mouse anti-Human, Mouse, Rat, Clone: T5P3G2/A3, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50195
This item is not returnable.
View return policy
Description
Cytochrome P450 26B1 Monoclonal specifically detects Cytochrome P450 26B1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Cytochrome P450 26B1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| CYP26A2, Cytochrome P450 26A2, cytochrome P450 26B1, cytochrome P450 CYP26B1 variant 2, Cytochrome P450 retinoic acid-inactivating 2, cytochrome P450 retinoid metabolizing protein, cytochrome P450, family 26, subfamily B, polypeptide 1, Cytochrome P450RAI-2, DKFZp686G0638, EC 1.14, MGC129613, P450RAI2, P450RAI-2, Retinoic acid-metabolizing cytochrome | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 56603 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| T5P3G2/A3 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| CYP26B1 | |
| Ovalbumin-conjugated synthetic peptide DSNQNEILPE (C-terminal sequence) | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction