missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 1A1/1A2 Antibody (MC1), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
£169.00
Specifications
| Antigen | Cytochrome P450 1A1/1A2 |
|---|---|
| Clone | MC1 |
| Dilution | Western Blot 1:500, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:20 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Cytochrome P450 1A1/1A2 Monoclonal specifically detects Cytochrome P450 1A1/1A2 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Cytochrome P450 1A1/1A2 | |
| Western Blot 1:500, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:20 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| AHH, AHRR, aryl hydrocarbon hydroxylase, CP11, CYP1, CYPIA1, cytochrome P1-450, dioxin-inducible, cytochrome P450 1A1, Cytochrome P450 form 6, cytochrome P450, family 1, subfamily A, polypeptide 1, Cytochrome P450-C, Cytochrome P450-P1, EC 1.14.14.1, flavoprotein-linked monooxygenase, P1-450, P450-C, P450DX, subfamily I (aromatic compound-inducible), polypeptide 1, xenobiotic monooxygenase | |
| CYP1A1 | |
| IgG1 | |
| Unpurified | |
| Detects Cytochrome P450 1A1 (Gene ID: 1543, UniProt P04798) and Cytochrom P450 1A2 (Gene ID: 1544, UniProt: P05177) from human samples. |
| MC1 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Breast Cancer, Lipid and Metabolism | |
| Ascites with 0.05% Sodium Azide | |
| 1543 | |
| 3-methylcholanthrene induced rat cytochrome P450 protein. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| 58 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title