missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 1A1/1A2 Antibody (MC1), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB100-74398-0.025ml
This item is not returnable.
View return policy
Description
Cytochrome P450 1A1/1A2 Monoclonal specifically detects Cytochrome P450 1A1/1A2 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Cytochrome P450 1A1/1A2 | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.05% Sodium Azide | |
| CYP1A1 | |
| 3-methylcholanthrene induced rat cytochrome P450 protein. | |
| Unpurified | |
| RUO | |
| Primary | |
| Detects Cytochrome P450 1A1 (Gene ID: 1543, UniProt P04798) and Cytochrom P450 1A2 (Gene ID: 1544, UniProt: P05177) from human samples. | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| MC1 | |
| Western Blot 1:500, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:20 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| AHH, AHRR, aryl hydrocarbon hydroxylase, CP11, CYP1, CYPIA1, cytochrome P1-450, dioxin-inducible, cytochrome P450 1A1, Cytochrome P450 form 6, cytochrome P450, family 1, subfamily A, polypeptide 1, Cytochrome P450-C, Cytochrome P450-P1, EC 1.14.14.1, flavoprotein-linked monooxygenase, P1-450, P450-C, P450DX, subfamily I (aromatic compound-inducible), polypeptide 1, xenobiotic monooxygenase | |
| Mouse | |
| 58 kDa | |
| 0.025 mL | |
| Breast Cancer, Lipid and Metabolism | |
| 1543 | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction