missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome b245 alpha Antibody (CS9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-40974-0.025MG
This item is not returnable.
View return policy
Description
Cytochrome b245 alpha Monoclonal specifically detects Cytochrome b245 alpha in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
| Cytochrome b245 alpha | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:100, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| P13498 | |
| CYBA | |
| Cytochrome B245 solubilised in octyl-beta-gluycopyranoside | |
| 0.025 mg | |
| Primary | |
| This antibody recognizes the light chain of cytochrome B245, also known as p22phox, a subunit of the nicotinamide adenine dinucleotide phosphate-oxidase (NADPH oxidase). NADPH oxidase is a membrane-bound enzyme complex in neutrophils that produces the superoxide anion (O2-) by transferring single electrons from NADPH inside the cell across the membrane and coupling them to molecular oxygen. Superoxide is used for the destruction of pathogens. Individuals with defects in NADPH oxidase suffer from chronic granulomatous disease, characterized by severe recurrent infections. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| ELISA, Immunohistochemistry (Frozen), Flow Cytometry, Immunoprecipitation, Western Blot, Immunohistochemistry | |
| CS9 | |
| Unconjugated | |
| PBS (pH 7.4) with 0.09% Sodium Azide | |
| cytochrome b light chain, Cytochrome b(558) alpha chain, cytochrome b(558) alpha-subunit, cytochrome b, alpha polypeptide, cytochrome b-245 light chain, cytochrome b-245, alpha polypeptide, Cytochrome b558 subunit alpha, flavocytochrome b-558 alpha polypeptide, Neutrophil cytochrome b 22 kDa polypeptide, p22 phagocyte B-cytochrome, p22phox, p22-phox, Superoxide-generating NADPH oxidase light chain subunit | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 1535 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction