missing translation for 'onlineSavingsMsg'
Learn More

Cytidine-5'-Triphosphate Disodium Salt Dihydrate

Product Code. 15855118 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Packaging:
Plastic bottle
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15855118 1 g Plastic bottle 1g
15825278 100 mg Plastic bottle 100mg
15835278 500 mg Plastic bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15855118 Supplier Thermo Scientific Supplier No. J14121.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 81012-87-5 | C9H18N3Na2O16P3 | 563.15 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81012-87-5
Molecular Formula C9H18N3Na2O16P3
Molecular Weight (g/mol) 563.15
MDL Number MFCD00150807
InChI Key KNSYTSUGTLJHPS-ODQFIEKDSA-L
Synonym cytidine 5 '-triphosphate sodium salt, cytidine 5 '-triphosphate tetrasodiμm
PubChem CID 129893565
IUPAC Name [[(2 R,3 S,4 R,5 R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] phosphono hydrogen phosphate;sodiμm
SMILES O.O.[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]1O

Specifications

Physical Form Powder
Quantity 1 g
Formula Weight 527.13 (anhydrous)
Packaging Plastic bottle
Chemical Name or Material Cytidine to 5 '-Triphosphate Disodium Salt Dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.