accessibility menu, dialog, popup

UserName

NH2NON(R)O(R)OPOOOPOOOPHOHOO(S)HO(R)OHNaNaH2OH2O

CAS RN 81012-87-5

IUPAC Name: ({[(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)(phosphonatooxy)phosphinate
Synonyms: ctp cytidine 5'-triphosphate(4-)
Molecular Weight (g/mol): 479.13
Molecular Formula: C9H12N3O14P3
InChi Key: PCDQPRRSZKQHHS-XVFCMESISA-J
SMILES: NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OP([O-])([O-])=O)[C@@H](O)[C@H]1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results