missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYTB Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£149.00 - £359.00
Specifications
| Antigen | CYTB |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18646168
|
Novus Biologicals
NBP3-05590-100ul |
100 μg |
£359.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18696008
|
Novus Biologicals
NBP3-05590-20ul |
20 μg |
£149.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CYTB Polyclonal antibody specifically detects CYTB in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| CYTB | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 4519 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| cytochrome b, MTCYB, MT-CYB mitochondrially encoded cytochrome b | |
| A synthetic peptide corresponding to a sequence within amino acids 3-103 of mouse CYTB (NP_904340.1). NMRKTHPLFKIINHSFIDLPAPSNISSWWNFGSLLGVCLMVQIITGLFLAMHYTSDTMTAFSSVTHICRDVNYGWLIRYMHANGASMFFICLFLHVGRGLY | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title