missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYTB Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05590-20ul
This item is not returnable.
View return policy
Description
CYTB Polyclonal antibody specifically detects CYTB in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| CYTB | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| cytochrome b, MTCYB, MT-CYB mitochondrially encoded cytochrome b | |
| A synthetic peptide corresponding to a sequence within amino acids 3-103 of mouse CYTB (NP_904340.1). NMRKTHPLFKIINHSFIDLPAPSNISSWWNFGSLLGVCLMVQIITGLFLAMHYTSDTMTAFSSVTHICRDVNYGWLIRYMHANGASMFFICLFLHVGRGLY | |
| 20 μg | |
| Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 4519 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction