Learn More
Agilent BioTek Cytation 1 Cell Imaging Multi-Mode Reader
BioTek Cytation 1 cell imaging multi-mode reader combines fluorescence and high contrast brightfield imaging with conventional multi-mode detection. For Research Use Only. Not for use in diagnostic procedures.
POA
Specifications
Type | Cell Imaging Multi-Mode Microplate Reader |
---|---|
Shaking | Linear, Orbital, Double Orbital |
CO2 Concentration Range | 0 to 20% |
O2 Concentration Range | 0 to 19% |
Plate Type | 6-well to 384-well plates |
Product Code | Brand | Model | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Includes | Price | Quantity & Availability | ||||
17267929
|
Agilent BioTek
CYT1V-SN |
CYT1V | Fluorescence and high contrast brightfield imaging |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
17247929
|
Agilent BioTek
CYT1FAV-SN |
CYT1FAV | Fluorescence and high contrast brightfield imaging; Monochromator absorbance; Filter/dichroic fluorescence; Luminescence/filtered luminescence |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
17227929
|
Agilent BioTek
CYT1FA-SN |
CYT1FA | Monochromator absorbance; Filter/dichroic fluorescence; Luminescence/filtered luminescence |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
Description
BioTek Cytation 1 cell imaging multi-mode reader combines fluorescence and high contrast brightfield imaging with conventional multi-mode detection. This affordable imaging reader delivers performance and economy compared to other digital microscopy systems
The BioTek Cytation 1 design provides both quantitative phenotypic cellular data with well-based quantitative data. The multi-mode detection module has high sensitivity filter-based fluorescence and a monochromator for UV-Vis absorbance. Temperature control and shaking are standard. CO2/O2 control and reagent injectors are available. Gen5 software simplifies image capture and plate reading.
Specifications
Cell Imaging Multi-Mode Microplate Reader | |
0 to 20% | |
6-well to 384-well plates | |
Xenon Flash | |
Gen5 Microplate Reader and Imager Software | |
20 in. | |
17.5 in. | |
16.5 in. | |
CE and TUV marked. RoHS compliant. |
Linear, Orbital, Double Orbital | |
0 to 19% | |
Endpoint, kinetic, area scanning, plate mode inject/read process and well mode inject/read process | |
0.3 μm/pixel at 20X | |
Incubation to 45°C | |
50.8 cm | |
44.5 cm | |
41.9 cm |