Learn More
Agilent BioTek Cytation 1 Cell Imaging Multi-Mode Reader
Shop All Agilent BioTek Products

Description
BioTek Cytation 1 cell imaging multi-mode reader combines fluorescence and high contrast brightfield imaging with conventional multi-mode detection. This affordable imaging reader delivers performance and economy compared to other digital microscopy systems
The BioTek Cytation 1 design provides both quantitative phenotypic cellular data with well-based quantitative data. The multi-mode detection module has high sensitivity filter-based fluorescence and a monochromator for UV-Vis absorbance. Temperature control and shaking are standard. CO2/O2 control and reagent injectors are available. Gen5 software simplifies image capture and plate reading.
Spécification
Spécification
| Type | Cell Imaging Multi-Mode Microplate Reader |
| Shaking | Linear, Orbital, Double Orbital |
| CO2 Concentration Range | 0 to 20% |
| O2 Concentration Range | 0 to 19% |
| Includes | Monochromator absorbance; Filter/dichroic fluorescence; Luminescence/filtered luminescence |
| Model | CYT1FA |
| Plate Type | 6-well to 384-well plates |
| Reading Options | Endpoint, kinetic, area scanning, plate mode inject/read process and well mode inject/read process |
| Light Source | Xenon Flash |
| Resolution | 0.3 μm/pixel at 20X |
| Afficher plus |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.