missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cystatin A Recombinant Rabbit Monoclonal Antibody (23GB5665), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552891
This item is not returnable.
View return policy
Description
The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins, and kininogens. This gene encodes a stefin that functions as a cysteine protease inhibitor, forming tight complexes with papain and the cathepsins B, H, and L. The protein is one of the precursor proteins of cornified cell envelope in keratinocytes and plays a role in epidermal development and maintenance. Stefins have been proposed as prognostic and diagnostic tools for cancer.
Specifications
| Cystatin A | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CSTA | |
| AREI; Csta; Csta1; cystatin A; cystatin A (stefin A); cystatin A1; cystatin AS; cystatin-A; Cystatin-A, N-terminally processed; Cystatin-A1; Cystatin-AS; PSS4; Stefin A; stefin-A; STF1; STFA | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 288067 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5665 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P01039 | |
| CSTA | |
| A synthesized peptide derived from human Cystatin A (75-98AA). | |
| 20 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction