missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cystatin A Recombinant Rabbit Monoclonal Antibody (23GB5665), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | Cystatin A |
|---|---|
| Clone | 23GB5665 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087893
![]() |
Invitrogen™
MA552890 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088889
|
Invitrogen™
MA552891 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cystatin A Recombinant Monoclonal antibody specifically detects Cystatin A in Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| Cystatin A | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Rat | |
| P01039 | |
| 288067 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5665 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AREI; Csta; Csta1; cystatin A; cystatin A (stefin A); cystatin A1; cystatin AS; cystatin-A; Cystatin-A, N-terminally processed; Cystatin-A1; Cystatin-AS; PSS4; Stefin A; stefin-A; STF1; STFA | |
| A synthesized peptide derived from human Cystatin A (75-98AA). | |
| Primary | |
| -20°C | |
| CSTA |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title