missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin-8 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92605-0.1ml
This item is not returnable.
View return policy
Description
Cystatin-8 Polyclonal antibody specifically detects Cystatin-8 in Mouse samples. It is validated for Western Blot
Specifications
| Cystatin-8 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| CRESCystatin-related epididymal spermatogenic protein, cystatin 8 (cystatin-related epididymal specific), cystatin-8, cystatin-related epididymal-specific | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 53-142 of human CST8 (NP_005483.1). MQEYNKESEDKYVFLVVKTLQAQLQVTNLLEYLIDVEIARSDCRKPLSTNEICAIQENSKLKRKLSCSFLVGALPWNGEFTVMEKKCEDA | |
| 0.1 mL | |
| Cell Biology | |
| 10047 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction