missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyr61/CCN1 Antibody (3A7.1B8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£175.00 - £381.00
Specifications
| Antigen | Cyr61/CCN1 |
|---|---|
| Clone | 3A7.1B8 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 4 ug/ml, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18195478
|
Novus Biologicals
NBP2-36490SS |
0.025 mg |
£175.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18719643
|
Novus Biologicals
NBP2-36490 |
0.1 mg |
£381.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyr61/CCN1 Monoclonal specifically detects Cyr61/CCN1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Cyr61/CCN1 | |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| O00622 | |
| 3491 | |
| Partial recombinant protein from human Cyr61/CCN1 protein (between residues 100-300). [Swiss-Prot O00622] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 3A7.1B8 | |
| Western Blot 4 ug/ml, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| CCN family member 1, CCN1IGF-binding protein 10, Cysteine-rich angiogenic inducer 61, cysteine-rich, angiogenic inducer, 61, GIG1cysteine-rich heparin-binding protein 61, IBP-10, IGFBP-10, IGFBP10cysteine-rich, anigogenic inducer, 61, Insulin-like growth factor-binding protein 10, protein CYR61, Protein GIG1 | |
| CCN1 | |
| IgG2b κ | |
| Protein G purified | |
| Immunogen's sequence similarity with other species: Rat (81%), Mouse (79%), Camel (88%), Naked Mole Rat (87%), Porcine (80%), Bovine/Sheep/Horse (82%), Chicken (63%) |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title