missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyr61/CCN1 Antibody (3A7.1B8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-36490
This item is not returnable.
View return policy
Description
Cyr61/CCN1 Monoclonal specifically detects Cyr61/CCN1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Cyr61/CCN1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 4 ug/ml, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
| O00622 | |
| CCN1 | |
| Partial recombinant protein from human Cyr61/CCN1 protein (between residues 100-300). [Swiss-Prot O00622] | |
| 0.1 mg | |
| Primary | |
| Immunogen's sequence similarity with other species: Rat (81%), Mouse (79%), Camel (88%), Naked Mole Rat (87%), Porcine (80%), Bovine/Sheep/Horse (82%), Chicken (63%) | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 3A7.1B8 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| CCN family member 1, CCN1IGF-binding protein 10, Cysteine-rich angiogenic inducer 61, cysteine-rich, angiogenic inducer, 61, GIG1cysteine-rich heparin-binding protein 61, IBP-10, IGFBP-10, IGFBP10cysteine-rich, anigogenic inducer, 61, Insulin-like growth factor-binding protein 10, protein CYR61, Protein GIG1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 3491 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction