missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP46A1 Mouse, anti-Human, Clone: 1E10D3, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CYP46A1 |
|---|---|
| Clone | 1E10D3 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18066205
|
Proteintech
68576-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18084315
|
Proteintech
68576-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CYP46A1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CYP46A1. This antibody recognizes human, mouse, rat, rabbit, pig antigen. The CYP46A1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| CYP46A1 | |
| 1000 μg/mL | |
| Liquid | |
| Human, Mouse, Rat, Rabbit, Pig | |
| CH24H, Cholesterol 24 hydroxylase, CP46, CYP46, CYP46A1, Cytochrome P450 46A1 | |
| Primary |
| 1E10D3 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Mouse | |
| 10858 | |
| IgG2b | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title