Learn More
CYP46A1 Mouse, anti-Human, Clone: 1E10D3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68576-1-IG-150UL
Description
The CYP46A1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CYP46A1. This antibody recognizes human, mouse, rat, rabbit, pig antigen. The CYP46A1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| CYP46A1 | |
| 1E10D3 | |
| CH24H, Cholesterol 24 hydroxylase, CP46, CYP46, CYP46A1, Cytochrome P450 46A1 | |
| 150 μL | |
| 10858 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, Immunohistochemistry, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Rabbit, Pig | |
| 150 kDa | |
| IgG2b |