missing translation for 'onlineSavingsMsg'
Learn More

CYP46A1 Antibody (N8-P6E4*H8) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

£150.00 - £315.00

Specifications

Antigen CYP46A1
Clone N8-P6E4*H8
Concentration 1 mg/mL
Dilution Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500
Applications Western Blot, ELISA, Immunohistochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18691148
View Documents
Novus Biologicals
NBP2-50194-0.025ml
0.025 mL
£150.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18618808
View Documents
Novus Biologicals
NBP2-50194
0.1 mL
£315.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CYP46A1 Monoclonal antibody specifically detects CYP46A1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications

Specifications

CYP46A1
1 mg/mL
Western Blot, ELISA, Immunohistochemistry
Unconjugated
Mouse
Lipid and Metabolism, Neuroscience
PBS
10858
IgG1
Protein A purified
N8-P6E4*H8
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500
Monoclonal
Purified
RUO
Human
CH24H, cholesterol 24-hydroxylase, CYP46CP46, Cytochrome P450 46A1, cytochrome P450, family 46, subfamily A, polypeptide 1, cytochrome P450, subfamily 46 (cholesterol 24-hydroxylase), EC 1.14.13.98
Ovalbumin-conjugated synthetic peptide- PVLCTLRPR (C-terminal sequence)
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.