missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP46A1 Antibody (N8-P6E4*H8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50194-0.025ml
This item is not returnable.
View return policy
Description
CYP46A1 Monoclonal antibody specifically detects CYP46A1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| CYP46A1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| CH24H, cholesterol 24-hydroxylase, CYP46CP46, Cytochrome P450 46A1, cytochrome P450, family 46, subfamily A, polypeptide 1, cytochrome P450, subfamily 46 (cholesterol 24-hydroxylase), EC 1.14.13.98 | |
| Ovalbumin-conjugated synthetic peptide- PVLCTLRPR (C-terminal sequence) | |
| 0.025 mL | |
| Lipid and Metabolism, Neuroscience | |
| 10858 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry | |
| N8-P6E4*H8 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction