missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP27C1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-56523
This item is not returnable.
View return policy
Description
CYP27C1 Polyclonal specifically detects CYP27C1 in Human samples. It is validated for Western Blot.
Specifications
| CYP27C1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| cytochrome P450 27C1, cytochrome P450, family 27, subfamily C, polypeptide 1, cytochrome P450, family 27, subfamily C, polypeptide 13, EC 1.14, FLJ16008 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 339761 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q4G0S4 | |
| CYP27C1 | |
| Synthetic peptides corresponding to CYP27C1(cytochrome P450, family 27, subfamily C, polypeptide 1) The peptide sequence was selected from the middle region of CYP27C1. Peptide sequence VTQEDLVIGGYLIPKGTQLALCHYATSYQDENFPRAKEFRPERWLRKGDL. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Canine: 85%; Rabbit: 85%; Chicken: 76%. | |
| Human, Bovine, Canine, Equine, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction