missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP24A1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | CYP24A1 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
CYP24A1 Polyclonal specifically detects CYP24A1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CYP24A1 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| 25-dihydroxyvitamin D(3) 24-hydroxylase, mitochondrial, CP241, CYP241,25-alphadihydroxyvitamin D3 24-hydroxylase, Cytochrome P450 24A1, cytochrome P450, family 24, subfamily A, polypeptide 1,24-OHase, cytochrome P450, subfamily XXIV (vitamin D 24-hydroxylase), Cytochrome P450-CC24, EC 1.14.13.n4, exo-mitochondrial protein, MGC126273, MGC126274, P450-CC24, vitamin D 24-hydroxylase, Vitamin D(3) 24-hydroxylase | |
| The immunogen is a synthetic peptide directed towards the C terminal region of human CYP24A1 (NP_000773). Peptide sequence LNTKVWQDHTLAWDTIFKSVKACIDNRLEKYSQQPSADFLCDIYHQNRLS | |
| Affinity purified |
| Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 1591 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title