missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP11A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
£290.00 - £459.00
Specifications
| Antigen | CYP11A1 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18477361
|
Novus Biologicals
NBP1-85368-25ul |
25 μL |
£290.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18206037
|
Novus Biologicals
NBP1-85368 |
0.1 mL |
£459.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CYP11A1 Polyclonal specifically detects CYP11A1 in Human, Rhesus Macaque samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| CYP11A1 | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 1583 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:RSVLVKGCQTFLSAPREGLGRLRVPTGEGAGISTRSPRPFNEIPSPGDNGWLNLYHFWRETGTHKVHLHHVQNFQKYGPIYREKLGNVESVYVIDPEDVALLFKSEGPNPERFLIPPWVAYHQYYQRPIGVLLKKSAA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Cholesterol desmolase, cholesterol monooxygenase (side-chain-cleaving), cholesterol side-chain cleavage enzyme, mitochondrial, CYPXIA1, Cytochrome P450 11A1, Cytochrome P450(scc), cytochrome P450, family 11, subfamily A, polypeptide 1, cytochrome P450C11A1, EC 1.14.15, EC 1.14.15.6, steroid 20-22-lyase, subfamily XIA (cholesterol side chain cleavage) | |
| CYP11A1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title