missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP11A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-85368
This item is not returnable.
View return policy
Description
CYP11A1 Polyclonal specifically detects CYP11A1 in Human, Rhesus Macaque samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| CYP11A1 | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Cholesterol desmolase, cholesterol monooxygenase (side-chain-cleaving), cholesterol side-chain cleavage enzyme, mitochondrial, CYPXIA1, Cytochrome P450 11A1, Cytochrome P450(scc), cytochrome P450, family 11, subfamily A, polypeptide 1, cytochrome P450C11A1, EC 1.14.15, EC 1.14.15.6, steroid 20-22-lyase, subfamily XIA (cholesterol side chain cleavage) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| CYP11A1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:RSVLVKGCQTFLSAPREGLGRLRVPTGEGAGISTRSPRPFNEIPSPGDNGWLNLYHFWRETGTHKVHLHHVQNFQKYGPIYREKLGNVESVYVIDPEDVALLFKSEGPNPERFLIPPWVAYHQYYQRPIGVLLKKSAA | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 1583 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction