missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CYM 50260

Product Code. 30001518
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001518 10 mg 10mg
30001536 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001518 Supplier Tocris Bioscience™ Supplier No. 4677/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective S1P4 agonist

Potent sphingosine-1-phosphate receptor 4 (S1P4) agonist (EC50 = 45 nM). Displays no activity at other S1P receptor subtypes (S1P1-3 and S1P5) at concentrations up to 25 μM.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1355026-60-6
Molecular Formula C14H11Cl3FNO2

Specifikationer

Quantity 10 mg
Formula Weight 350.6
Purity >98%
Molecular Weight (g/mol) 350.595
Chemical Name or Material CYM 50260
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.