CAS RN 1355026-60-6
CAS RN 1355026-60-6
IUPAC Name:
2-chloro-3-[2-(2,4-dichlorophenoxy)ethoxy]-6-(fluoromethyl)pyridine
Synonyms:
2-chloro-3-[2-(2,4-dichlorophenoxy)ethoxy]-6-(fluoromethyl)pyridine
Molecular Weight (g/mol):
350.6
Molecular Formula:
C14H11Cl3FNO2
InChi Key:
FHPOTBQOUBMMCI-UHFFFAOYSA-N
SMILES:
FCC1=NC(Cl)=C(OCCOC2=C(Cl)C=C(Cl)C=C2)C=C1
1
–
1
of
1
results
1
Promotions Available
Tocris Bioscience™ CYM 50260
Tocris Bioscience™ CYM 50260
CAS: 1355026-60-6 Molecular Formula: C14H11Cl3FNO2