missing translation for 'onlineSavingsMsg'
Learn More

GE Healthcare CyDye DIGE Fluor Cy2 Minimal Dye

Product Code. 11700455 Shop All Cytiva Products
Click to view available options
Quantity:
25 nmol
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11700455

Brand: GE Healthcare 25190027

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Formulation 100% Cy2 (Bis NHS ester)
For Use With (Application) Suitable for larger experimental set-ups with several variables, up to 60 labelings
Quantity 25 nmol
Product Type CyDye DIGE Fluor Cy2 Minimal Dye
Kit Contents 25 nmol (5 x 5 nmol) of CyDye DIGE fluor Cy2 minimal dye
Content And Storage -15°C to -30°C
Label or Dye Cy2
Color Blue
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.