missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclophilin-E Mouse anti-Human, Clone: 1.70E+09, Novus Biologicals™
Mouse Monoclonal Antibody
£234.00 - £378.00
Specifications
| Antigen | Cyclophilin-E |
|---|---|
| Clone | 1700000000 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, ELISA |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18262482
|
Novus Biologicals
NBP2-59502-50UL |
50 μL |
£234.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18280165
|
Novus Biologicals
NBP2-59502-100UL |
100 μL |
£378.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclophilin-E Monoclonal specifically detects Cyclophilin-E in Human samples. It is validated for Western Blot, ELISA.Specifications
| Cyclophilin-E | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| Cyclophilin E, Cyclophilin-33, CyP-33, CYP33, EC 5.2.1.8, MGC111222, MGC3736, peptidyl-prolyl cis-trans isomerase E, peptidylprolyl isomerase E (cyclophilin E), PPIase E, Rotamase E | |
| PPIE | |
| IgG2b κ | |
| Protein A purified |
| 1700000000 | |
| Western Blot 1:1000, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| 10450 | |
| Recombinant human PPIE (1-301aa) purified from E. coli | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title