missing translation for 'onlineSavingsMsg'
Learn More

Cyclophilin-E Mouse anti-Human, Clone: 1.70E+09, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-59502-50UL

 View more versions of this product

Product Code. 18262482

  • £234.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Cyclophilin-E Monoclonal specifically detects Cyclophilin-E in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cyclophilin-E
Monoclonal
1 mg/mL
Western Blot 1:1000, ELISA
Cyclophilin E, Cyclophilin-33, CyP-33, CYP33, EC 5.2.1.8, MGC111222, MGC3736, peptidyl-prolyl cis-trans isomerase E, peptidylprolyl isomerase E (cyclophilin E), PPIase E, Rotamase E
Mouse
Protein A purified
RUO
10450
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG2b κ
Western Blot, ELISA
1700000000
Unconjugated
Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide
PPIE
Recombinant human PPIE (1-301aa) purified from E. coli
50 μL
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only