missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Cyclones

Product Code. 12652705 Shop All Retsch Products
Change view
Click to view available options
Includes:
250, 500 mL Sample Bottles, Filter Bag
250, 500 mL Sample Bottles, Filter Bag, Connection for Vacuum Cleaner
3 L Collecting Receptacle, Filter Bag
3 L Collecting Receptacle, Filter Bag, Connection for Vacuum Cleaner
5 L Collecting Receptacle, Filter Bag, Connection for Vacuum Cleaner
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
12652705 3 L Collecting Receptacle, Filter Bag, Connection for Vacuum Cleaner Each
11737404 250, 500 mL Sample Bottles, Filter Bag Each
12642705 3 L Collecting Receptacle, Filter Bag Each
11984272 250, 500 mL Sample Bottles, Filter Bag, Connection for Vacuum Cleaner Each
11994272 5 L Collecting Receptacle, Filter Bag, Connection for Vacuum Cleaner Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 12652705 Supplier RETSCH Supplier No. 22.935.0018
  • ONLINE EXCLUSIVE PRICE
    £2680.00
    £2550.00 / Pack of 1
    Save £130.00
    4% Off
Estimated Shipment: 21-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Product Type Cyclone
Diameter (Metric) 31.2 mm
For Use With (Equipment) For ZM 200
Includes 3 L Collecting Receptacle, Filter Bag, Connection for Vacuum Cleaner
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.