missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanecarboxaldehyde, 97%

Product Code. 10366740 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10366740 100 mL Glass bottle 100mL
10592731 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10366740 Supplier Thermo Scientific Acros Supplier No. 148671000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2043-61-0 | C7H12O | 112.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2043-61-0
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00001457
InChI Key KVFDZFBHBWTVID-UHFFFAOYSA-N
Synonym cyclohexanecarboxaldehyde, cyclohexanal, formylcyclohexane, cyclohexanealdehyde, cyclohexanaldehyde, cyclohexylcarboxaldehyde, 1-formylcyclohexane, hexahydrobenzaldehyde, cyclohexylformaldehyde, cyclohexane formaldehyde
PubChem CID 16275
IUPAC Name cyclohexanecarbaldehyde
SMILES O=CC1CCCCC1

Specifications

Color Colorless to Pink
Density 0.9260g/mL
Boiling Point 161.0°C to 163.0°C
Flash Point 41°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H11CHO
Refractive Index 1.4490 to 1.4520
Quantity 100 mL
Beilstein 07, 19
Specific Gravity 0.926
Formula Weight 112.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Cyclohexanecarboxaldehyde, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cyclohexanecarboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.