accessibility menu, dialog, popup

UserName

O

CAS RN 2043-61-0

IUPAC Name: cyclohexanecarbaldehyde
Synonyms: cyclohexanecarboxaldehyde
Molecular Weight (g/mol): 112.17
Molecular Formula: C7H12O
InChi Key: KVFDZFBHBWTVID-UHFFFAOYSA-N
SMILES: O=CC1CCCCC1
Boiling Point 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)