missing translation for 'onlineSavingsMsg'
Learn More

Cycloheptanecarbonitrile, 98%

Product Code. 11475510
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11475510 5 g 5g
11485510 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11475510 Supplier Thermo Scientific Alfa Aesar Supplier No. H25804.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 32730-85-1 | C8H13N | 123.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32730-85-1
Molecular Formula C8H13N
Molecular Weight (g/mol) 123.20
MDL Number MFCD00004148
InChI Key UIZRDFICGGONJJ-UHFFFAOYSA-N
Synonym cycloheptyl cyanide, cycloheptylcyanide, cycloheptankohlenitrile, acmc-20ald5
PubChem CID 96653
IUPAC Name cycloheptanecarbonitrile
SMILES N#CC1CCCCCC1

Specifications

Density 0.925
Boiling Point 85°C to 86°C (10 mmHg)
Flash Point 85°C (185°F)
Refractive Index 1.464
Quantity 5 g
UN Number UN3276
Formula Weight 123.2
Percent Purity 98%
Chemical Name or Material Cycloheptanecarbonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.