CAS RN 32730-85-1
IUPAC Name:
cycloheptanecarbonitrile
Synonyms:
cycloheptanecarbonitrile
Molecular Weight (g/mol):
123.2
Molecular Formula:
C8H13N
InChi Key:
UIZRDFICGGONJJ-UHFFFAOYSA-N
SMILES:
N#CC1CCCCCC1
1
–
1
of
1
results
1
Cycloheptanecarbonitrile, 98%
CAS: 32730-85-1 Molecular Formula: C8H13N Molecular Weight (g/mol): 123.20 MDL Number: MFCD00004148 InChI Key: UIZRDFICGGONJJ-UHFFFAOYSA-N Synonym: cycloheptyl cyanide,cycloheptylcyanide,cycloheptankohlenitrile,acmc-20ald5 PubChem CID: 96653 IUPAC Name: cycloheptanecarbonitrile SMILES: N#CC1CCCCCC1