missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutyl methyl ketone, 97%

Product Code. p-7043930
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11352196

Brand: Thermo Scientific Alfa Aesar L03521.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3019-25-8 | C6H10O | 98.145 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3019-25-8
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00037148
InChI Key JPJOOTWNILDNAW-UHFFFAOYSA-N
Synonym cyclobutyl methyl ketone, 1-cyclobutylethan-1-one, acetylcyclobutane, ethanone, 1-cyclobutyl, ketone, cyclobutyl methyl, methyl cyclobutyl ketone, cyclobutylmethylketone, acetylcyclobutan, cyclobutylethanone, cyclobutane, acetyl
PubChem CID 76398
IUPAC Name 1-cyclobutylethanone
SMILES CC(=O)C1CCC1

Specifications

Density 0.9
Boiling Point 134°C to 135°C
Flash Point 28°C (82°F)
Refractive Index 1.432
Quantity 1 g
UN Number UN1224
Beilstein 1924022
Formula Weight 98.15
Percent Purity 97%
Chemical Name or Material Cyclobutyl methyl ketone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.