CAS RN 3019-25-8
CAS RN 3019-25-8
IUPAC Name:
1-cyclobutylethan-1-one
Synonyms:
ethanone, 1-cyclobutyl-
Molecular Weight (g/mol):
98.15
Molecular Formula:
C6H10O
InChi Key:
JPJOOTWNILDNAW-UHFFFAOYSA-N
SMILES:
CC(=O)C1CCC1
1
–
1
of
1
results
1
Cyclobutyl methyl ketone, 97%
CAS: 3019-25-8 Molecular Formula: C6H10O Molecular Weight (g/mol): 98.145 MDL Number: MFCD00037148 InChI Key: JPJOOTWNILDNAW-UHFFFAOYSA-N Synonym: cyclobutyl methyl ketone,1-cyclobutylethan-1-one,acetylcyclobutane,ethanone, 1-cyclobutyl,ketone, cyclobutyl methyl,methyl cyclobutyl ketone,cyclobutylmethylketone,acetylcyclobutan,cyclobutylethanone,cyclobutane, acetyl PubChem CID: 76398 IUPAC Name: 1-cyclobutylethanone SMILES: CC(=O)C1CCC1