missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutanol, 99+%

Product Code. 10307830 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10307830 1 g Glass bottle 1g
10635802 250 mg Glass bottle 250mg
10637104 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10307830 Supplier Thermo Scientific Acros Supplier No. 168750010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2919-23-5 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2919-23-5
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00001318
InChI Key KTHXBEHDVMTNOH-UHFFFAOYSA-N
Synonym cyclobutyl alcohol, cyclobutyl hydroxide, hydroxycyclobutane, acmc-1cgim, cyclobutanol, cyclobutanol 1g, ksc206s6t, kthxbehdvmtnoh-uhfffaoysa
PubChem CID 76218
IUPAC Name cyclobutanol
SMILES OC1CCC1

Specifications

Color Colorless
Density 0.9200g/mL
Boiling Point 122.0°C to 123.0°C (733.0 mmHg)
Flash Point 21°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C4H7OH
Refractive Index 1.4350 to 1.4370
Quantity 1 g
Beilstein 06, 4
Specific Gravity 0.92
Formula Weight 72.11
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Cyclobutanol
Show More Show Less
compliance-icons
Product Identifier
  • Cyclobutanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.