accessibility menu, dialog, popup

UserName

CAS RN 2919-23-5

IUPAC Name: cyclobutanol
Synonyms: cyclobutanol
Molecular Weight (g/mol): 72.11
Molecular Formula: C4H8O
InChi Key: KTHXBEHDVMTNOH-UHFFFAOYSA-N
SMILES: OC1CCC1
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)