missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutanol, 96%

Product Code. 11479633
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479633 5 g 5g
11469633 1 g 1g
11489633 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479633 Supplier Thermo Scientific Alfa Aesar Supplier No. A12430.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2919-23-5 | C4H8O | 72.11 g/mol

Has synthetic applications of gold-catalyzed ring expansions. Development of novel type of transition metal-catalyzed cascade reactions utilizing small ring systems and the applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Has synthetic applications of gold-catalyzed ring expansions. Development of novel type of transition metal-catalyzed cascade reactions utilizing small ring systems and the applications.

Solubility
Partly soluble.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acid chlorides, acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2919-23-5
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00001318
InChI Key KTHXBEHDVMTNOH-UHFFFAOYSA-N
Synonym cyclobutyl alcohol, cyclobutyl hydroxide, hydroxycyclobutane, acmc-1cgim, cyclobutanol, cyclobutanol 1g, ksc206s6t, kthxbehdvmtnoh-uhfffaoysa
PubChem CID 76218
IUPAC Name cyclobutanol
SMILES OC1CCC1

Specifications

Density 0.925
Boiling Point 123°C to 124°C
Flash Point 21°C (69°F)
Refractive Index 1.436
Quantity 5 g
UN Number UN1120
Beilstein 2035937
Solubility Information Partly soluble.
Formula Weight 72.11
Percent Purity 96%
Chemical Name or Material Cyclobutanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.